Learn More
Medchemexpress LLC N-Acetyl-L-tryptophan (Standard) | 1218-34-4 | 99.99% | 246.26 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW011978R
Encompass_Preferred
N-Acetyl-L-tryptophan (Standard) is an analytical standard and endogenous metabolite intended for research and analytical applications. It serves as a reference for assays and is utilized in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Functions as an analytical standard
  • Serves as a reference for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Functions as an endogenous metabolite
Catalog No. 50-004-51375