Learn More
Medchemexpress LLC N-acetyl-L-tryptophan (standard) | 1218-34-4 | MFCD00065976 | 99.99% | 246.26 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW011978R
N-Acetyl-L-tryptophan (Standard) is the analytical standard of N-Acetyl-L-tryptophan. This product is intended for research and analytical applications. N-Acetyl-L-tryptophan is an endogenous metabolite. For research use only.
- The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
- Appearance: solid
- Color: white to off-white
- Initial source: endogenous metabolite
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: please store the product under the recommended conditions in the certificate of analysis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.