Learn More
Cayman Chemical N-Acetyl-L-tyrosIn 100g
Supplier: Cayman Chemical 34345100 G
An active metabolite of, and precursor to, L-tyrosine; increases B. mori larvae heat stress tolerance; increases A. mellifera survival in a model of environmental stress; blood levels are increased during heat and restraint stress in mice; dietary administration decreases blood lipid peroxidation and corticosterone levels in a mouse model of restraint stress; dietary administration decreases tumor volume in an HCT116 mouse xenograft model; urinary excretion is increased in patients with PKU or tyrosinemia