Learn More
Medchemexpress LLC N-acetyl mesalazine | 51-59-2 | 99.9% | C9H9NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY6600810MM/1ML
Encompass_Preferred
N-acetyl mesalazine is the primary intestinal metabolite of 5-aminosalicylic acid and acts as a biomarker for its efficacy. It can scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation. This compound is suitable for research into conditions such as colitis and colon cancer.
  • Primary intestinal metabolite of 5-aminosalicylic acid
  • Serves as a biomarker for 5-aminosalicylic acid efficacy
  • Scavenges free radicals
  • Reduces DNA base hydroxylation
  • Ameliorates mucosal inflammation
  • Suitable for research into colitis and colon cancer
  • Inhibits •OH radical-stimulated DNA base hydroxylation in calf thymus DNA
Catalog No. 50-004-33584