Learn More
Medchemexpress LLC N-Acetyl mesalazine | 51-59-2 | 99.9% | C9H9NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY66008100MG
Encompass_Preferred
N-Acetyl mesalazine (N-Acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-Aminosalicylic Acid and serves as a biomarker for evaluating the efficacy of 5-Aminosalicylic Acid. It can scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation. N-Acetyl mesalazine can be used in the study of diseases such as colitis and colon cancer.
  • Primary intestinal metabolite of 5-Aminosalicylic Acid.
  • Serves as a biomarker for evaluating the efficacy of 5-Aminosalicylic Acid.
  • Scavenges free radicals.
  • Reduces DNA base hydroxylation.
  • Ameliorates mucosal inflammation.
  • Can be used in the study of diseases such as colitis and colon cancer.
Catalog No. 50-004-33815