Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Acetyl mesalazine | 51-59-2 | 99.9% | C9H9NO4 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY66008

Encompass_Preferred

N-Acetyl mesalazine (N-Acetyl-5-aminosalicylic acid) is the primary intestinal metabolite of 5-Aminosalicylic Acid. It serves as a biomarker for evaluating the efficacy of 5-Aminosalicylic Acid and can scavenge free radicals, reduce DNA base hydroxylation, and ameliorate mucosal inflammation. This compound is used in the study of diseases such as colitis and colon cancer.

  • Solid appearance
  • Light brown to brown color
  • Molecular weight of 195.17
  • Inhibits •OH radical-stimulated DNA base hydroxylation
  • Store at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light

Catalog No. 50-003-59216


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.