Learn More
Cayman Chemical N-Acetyl-S-alyl-L-cysteIn 10mg
SDP
Supplier:  Cayman Chemical 1759210 MG
Encompass_Preferred
A principal metabolite of L-deoxyalliin in humans, mice, rats, and dogs that is readily detected in plasma and urine
Catalog No. 50-296-5665