Learn More
TARGETMOL CHEMICALS INC N-Acetyl-Ser-Asp-Lys-Pro 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TP182810MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Acetyl Ser-Asp-Lys-Pro is formed in bone marrow cells by enzymatic processing of thymosin beta4. It inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. Purity 99.85%
Catalog No. 50-288-0420