Learn More
Medchemexpress LLC N-Acetyl-Ser-dAsp-Lys-Pro acetate | 99.88% | 547.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0266B
Encompass_Preferred
N-Acetyl-Ser-dAsp-Lys-Pro (Ac-SdKP) acetate acts as a specific substrate for the N-terminal active site of angiotensin-converting enzyme (ACE). This compound is also recognized as a natural inhibitor of pluripotent hematopoietic stem cell proliferation. It possesses both anti-inflammatory and antifibrotic properties.
  • Specific substrate for the N-terminal active site of angiotensin-converting enzyme (ACE).
  • Natural inhibitor of pluripotent hematopoietic stem cell proliferation.
  • Exhibits anti-inflammatory properties.
  • Demonstrates antifibrotic properties.
Catalog No. 50-004-26003