Learn More
TARGETMOL CHEMICALS INC N-ACETYLCYSTEINE AMIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T551825MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. N-Acetylcysteine amide is a cell membranes and blood brain barrier permeant thiol antioxidant and neuroprotective agentpurity: 99%
Catalog No. 50-257-9752