Learn More
Matrix Scientific N-Acetyl-L-cysteine, 616-91-1, MFCD00004880, 100g
Supplier: Matrix Scientific 075227100G
Molecular Formula C5H9NO3S, Purity 95+%, Molecular Weight 163.20, Melting Point 106-111°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.