Learn More
Medchemexpress LLC N-acetylglycine | 543-24-8 | MFCD00004275 | 99.8% | 117.10 | C4H7NO3 | 5g
SDP
Supplier:  Medchemexpress LLC HYY00695G
Encompass_Preferred
N-Acetylglycine (Aceturic acid) is a minor constituent of numerous foods N-Acetylglycine is a flavor enhancer which elicits Monosodium glutamate (MSG)-like sensory experiencesN-Acetylglycine[1]
Catalog No. 50-002-68980