Learn More
Medchemexpress LLC N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 2 (human recombinant) | 10 UG

Supplier: Medchemexpress LLC HYP76160A10UG
Recombinant human B3GNT2 (beta-1,3-N-acetylglucosaminyltransferase 2) produced in HEK293 cells and supplied as a lyophilized powder. The protein carries a His tag, is glycosylated, and displays heterogeneous molecular weight by SDS-PAGE. Manufacturer datasheet provides formulation, reconstitution, and storage guidance for research use only.
- Human recombinant protein expressed in HEK293 cells.
- C-terminal His tag to facilitate detection and purification.
- Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
- Observed molecular weight ~55-130 kDa due to glycosylation.
- Reconstitution recommended at ≥100 μg/mL; avoid lower concentrations.
- Store at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.
- After reconstitution, stable at 4°C for 1 week or longer at -20°C with carrier protein.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.