Learn More
Research Products International Corp NANA [n-Acetylneuraminic acid, Synthetic Sialic Acid], 100 Milligrams
SDP
Supplier:  Research Products International Corp A100800.1
Encompass_Preferred
Substrate for N-Acetylneuraminic acid aldolase.
Catalog No. 50-213-105