Learn More
Medchemexpress LLC N-Acetylornithine | 6205-08-9 | 98.0% | 174.20 | 1 ML
Supplier: Medchemexpress LLC HY11308010MM/1ML
N-Acetylornithine is a precursor of ornithine, associated with type 2 diabetes. It is hydrolyzed by N-acetyl-L-ornithine deacetylase to produce ornithine and acetic acid.
- Precursor of ornithine
- Associated with type 2 diabetes
- Hydrolyzed by N-acetyl-L-ornithine deacetylase