Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Acetylpenicillamine | 15537-71-0 | ≥98.0% | 191.25 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0124985G

Encompass_Preferred

N-Acetylpenicillamine is a chemical compound used as a laboratory chemical for the manufacture of substances. It has a molecular weight of 191.25 and appears as a solid. This compound exhibits a melting point range of 185-190 °C and a boiling point of approximately 392.7 °C. It is important to note that N-Acetylpenicillamine is classified with hazards including skin and eye irritation, and potential respiratory irritation.

  • Solid appearance
  • Melting point range of 185-190 °C
  • Boiling point of 392.7±37.0 °C at 760 mmHg
  • Recommended powder storage at -20 °C for 3 years or 4 °C for 2 years
  • Recommended in-solvent storage at -80 °C for 6 months or -20 °C for 1 month

Catalog No. 50-004-53026


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.