Learn More
Medchemexpress LLC N-Acetylprocainamide | 32795-44-1 | MFCD00009052 | 99.89% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB110950MG
Encompass_Preferred
N-Acetylprocainamide, also known as Acecainide, is identified as a class III antiarrhythmic compound that functions by blocking potassium (K+) channels. It is supplied as a solid, appearing white to off-white, and has a molecular weight of 277.36 with the formula C15H23N3O2. The product is strictly for research purposes and not for human use.
  • Functions as a class III antiarrhythmic.
  • Blocks potassium channels.
  • Available in both solid and solution forms for various experimental needs.
  • High purity of 99.89% ensures reliable research outcomes.
  • Detailed solubility information and preparation protocols are provided for both in vitro and in vivo applications.
Catalog No. 50-004-13484