Learn More
Cayman Chemical N-AcetylpuromycIn 10mg
SDP
Supplier:  Cayman Chemical 2533510 MG
Encompass_Preferred
A non-ribotoxic form of puromycin; formed in puromycin-resistant S. alboniger that endogenously express puromycin-acetyltransferase
Catalog No. 50-293-4217