Learn More
Medchemexpress LLC N-Acetylputrescine hydrochloride | 18233-70-0 | 99.0% | 166.65 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113100
Encompass_Preferred
N-Acetylputrescine hydrochloride is a polyamine metabolite that regulates cell proliferation and signal transduction by participating in polyamine acetylation modification. It may reduce the positive charge of polyamines, affect protein-nucleic acid interactions, and promote cell growth. Its concentration changes can assist in disease diagnosis and microbial cell wall structure research.
  • Regulates cell proliferation and signal transduction
  • Participates in polyamine acetylation modification
  • May reduce the positive charge of polyamines
  • Affects protein-nucleic acid interactions
  • Promotes cell growth
  • Potential biomarker for lung cancer progression
  • Potential biomarker for Parkinson's disease
Catalog No. 50-004-39079