Learn More
Medchemexpress LLC N-acetyltryptamine | 1016-47-3 | MFCD00209910 | 96.4% | 202.25 g/mol | C12H14N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10090825MG
Encompass_Preferred
N-acetyltryptamine is an acetylated tryptamine derivative used as a research reagent. It acts as a partial agonist at melatonin receptors with reported activity in the retina, and is applied in assays such as determination of serotonin N-acetyl transferase activity. Supplied as a powder for laboratory research; intended for research use only.
  • Partial agonist at melatonin receptors
  • Applicable to assays of serotonin N-acetyl transferase activity
  • Powder form with defined storage recommendations (short- and long-term)
  • High chemical purity (96.4%)
  • CAS 1016-47-3; molecular weight 202.25 g/mol
  • Intended for research use only
Catalog No. 50-002-12237