Learn More
Cayman Chemical N aLinolenoyl Tyrosine
SDP
Supplier:  Cayman Chemical 1003210 MG
Encompass_Preferred
A simple α-amide conjugate between the ω-3 essential fatty acid α-linolenate and the amino acid tyrosine that was prepared as a mechanism for enhancing CNS dopamine content by facilitated transport of the tyrosine precursor across the blood-brain barrier; increased CNS dopamine levels and exhibited an activity profile consistent with an anti-Parkinson’s therapeutic agent in experimental rat models of dopamine insufficiency
Catalog No. 50-288-9697