Learn More
Medchemexpress LLC N-alkylated tranylcypromine | 2262489-89-2 | 98.1% | 437.91 g·mol⁻1 | C22H26ClF2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1365225MG
Encompass_Preferred
S2116 is a research-grade N-alkylated tranylcypromine derivative that acts as a potent lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K9 methylation and can induce apoptosis in cellular models, making it suitable for epigenetic and mechanistic studies in the laboratory.
  • Potent LSD1 inhibition with demonstrated cellular activity.
  • Increases H3K9 methylation and modulates histone marks.
  • High purity (~98.1%) suitable for biochemical assays.
  • Soluble in DMSO at 100 mg/mL (approx. 228 mM).
  • Available in small research pack sizes; larger quantities by request.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-22819