Learn More
Medchemexpress LLC N-alpha-(9-fluorenylmethoxycarbonyl)-N-zeta-(tert-butoxycarbonyl)-homo-L-lysine | 194718-17-7 | MFCD01860606 | 98.7% | 482.57 | C27H34N2O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1419851G
Encompass_Preferred
Fmoc-hLys(Boc)-OH is a Fmoc- and Boc-protected homolysine derivative used as a building block in Fmoc-based solid-phase peptide synthesis. The α-amino group is protected with an Fmoc group and the ε-side chain with a Boc group, enabling orthogonal deprotection and controlled incorporation into peptide sequences.
  • Protected amino acid for solid-phase peptide synthesis.
  • Enables selective side-chain protection and deprotection.
  • Compatible with standard Fmoc coupling and deprotection chemistries.
  • Supplied as a stable solid powder for synthesis workflows.
  • High purity suitable for research use (batch purity ~98.7%).
  • Available in multiple pack sizes for small-scale and bulk synthesis.
Catalog No. 50-002-83731