Learn More
Matrix Scientific N-alpha-methyl-L-proline, 475-11-6, MFCD00011565, 1g
Supplier: Matrix Scientific 0424861G
Molecular Formula C6H11NO2, Purity 95%, Molecular Weight 129.16, Melting Point 114-116°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.