Learn More
Cayman Chemical N-Alyl- -SKF 38393hydrob 5mg
SDP
Supplier:  Cayman Chemical 327765 MG
Encompass_Preferred
A dopamine D1 receptor partial agonist; selective for dopamine D1 over D2 receptors in radioligand binding assays (Kis = 302 and 5,250 nM, respectively); reduces food intake in rats (ED50 = 3.6 mg/kg); reduces cocaine and food self-administration in squirrel monkeys from 0.1-10 mg/kg
Catalog No. 50-295-0193