Learn More
Medchemexpress LLC N-(Amino-PEG1)-N-bis(PEG2-propargyl) | 2100306-47-4 | 98.0% | 356.46 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140086100MG
Encompass_Preferred
N-(Amino-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. This compound also functions as a click chemistry reagent, containing an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide group-containing molecules
Catalog No. 50-003-49767