Learn More
Medchemexpress LLC N-(Amino-PEG1)-N-bis(PEG2-propargyl) | 2100306-47-4 | 98.0% | 356.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140086
Encompass_Preferred
N-(Amino-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-50138