Learn More
Enzo Life Sciences N-Arachidonoyl-serotonin (50 mg). CAS: 187947-37-1
SDP
Supplier:  Enzo Life Sciences BMLVR1060050
Encompass_Preferred
Unnatural fatty acid-neurotransmitter conjugate. Potent TRPV1 antagonist (IC50=40nM). Inhibits fatty acid amide hydrolase (FAAH) (IC50=5.6mM). Enhances levels of anandamide and 2-AG in vivo. Displays analgesic activity in both acute and chronic peripheral pain models but shows little efficacy in an inflammatory bowel disease model. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol (15mg/ml) or dimethyl formamide (30mg/ml); also soluble in PBS, pH 7.2 (290µg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2356