Learn More
Cayman Chemical N-ArachIdonyl DopamIn 100mg
SDP
Supplier:  Cayman Chemical 90057100 MG
Encompass_Preferred
An arachidonoyl amino acid and CB1 receptor agonist (Ki = 250 nM in rat brain membranes, which highly express CB1 receptors); selective for CB1 over CB2 receptors (Ki = 12,000 nM in rat spleen membranes, which highly express CB2 receptors); induces intracellular calcium mobilization in N18TG2 neuroblastoma cells (EC50 = 0.7 µM); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 0.25 µM); induces hypothermia, catalepsy, hypolocomotion, and analgesia in mice at 10 mg/kg
Catalog No. 50-289-5810