Learn More
Cayman Chemical N-ArachIdonyl TaurIn-d4 500ug
SDP
Supplier:  Cayman Chemical 26323500 UG
Encompass_Preferred
An internal standard for the quantification of N-arachidonoyl taurine by GC- or LC-MS
Catalog No. 50-293-5257