Learn More
Medchemexpress LLC N-(azido-PEG2)-N-Boc-PEG3-NHS ester | 2093153-85-4 | 97.0% | C24H41N5O11 | 100 MG
Supplier: Medchemexpress LLC HY140559100MG
N-(Azido-PEG2)-N-Boc-PEG3-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
- PEG-based PROTAC linker
- Used in PROTAC synthesis
- Functions as a click chemistry reagent
- Contains an Azide group
- Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
- Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)