Learn More
Medchemexpress LLC N-(Azido-PEG2)-N-Boc-PEG4-NHS ester | 2093153-95-6 | C26H45N5O12 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140560
Encompass_Preferred
N-(Azido-PEG2)-N-Boc-PEG4-NHS ester is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It acts as a click chemistry reagent, facilitating reactions with molecules containing Alkyne, DBCO, or BCN groups through its Azide functionality. This product is designed for research applications.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Intended for research use only
Catalog No. 50-003-52738