Learn More
Medchemexpress LLC N-(Azido-PEG2)-N-Boc-PEG4-NHS ester | 2093153-95-6 | Pure form | C26H45N5O12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140560100MG
Encompass_Preferred
N-(Azido-PEG2)-N-Boc-PEG4-NHS ester is a versatile PEG-based PROTAC linker. It is commonly employed in the synthesis of PROTACs, which are bifunctional molecules designed to induce targeted protein degradation. This compound also functions as a click chemistry reagent, featuring an Azide group. This Azide group enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-53091