Learn More
Medchemexpress LLC N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester | 2100306-76-9 | 95.0% | C30H54N6O11S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140581
Encompass_Preferred
N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a click chemistry reagent
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilized in the synthesis of PROTACs
Catalog No. 50-003-52736