Learn More
Medchemexpress LLC N-(Azido-PEG4)-biocytin | 2055042-70-9 | C27H47N7O9S | 250MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140919
Encompass_Preferred
N-(Azido-PEG4)-biocytin is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group-containing molecules.
  • Used in the synthesis of PROTACs
  • Click chemistry reagent containing an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Reacts with alkyne-containing molecules
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Reacts with DBCO or BCN group-containing molecules
  • PROTACs utilize the ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-003-52877