Learn More
Medchemexpress LLC N-(Azido-PEG4)-biocytin | 2055042-70-9 | C27H47N7O9S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14091950MG
Encompass_Preferred
N-(Azido-PEG4)-biocytin is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group for CuAAc reactions
  • Facilitates SPAAC reactions with DBCO or BCN groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55075