Learn More
Medchemexpress LLC N-(benzo[d]thiazol-6-yl)-5-(4-hydroxyphenyl)furan-2-carboxamide | 2285432-57-5 | 98.2% | 336.36 g·mol⁻¹ | C18H12N2O3S | 10 MG

Supplier: Medchemexpress LLC HY15389510MG
SLU-PP-1072 is a small-molecule dual inverse agonist of estrogen-related receptors ERRα and ERRγ used in preclinical prostate cancer research. It disrupts cancer cell metabolism, inhibits the Warburg effect, and induces apoptosis through dysregulation of the cell cycle. Intended for in vitro research use only.
- Dual ERRα/γ inverse agonist.
- Disrupts prostate cancer cell metabolism and the Warburg effect.
- Induces apoptosis via cell cycle dysregulation.
- Suitable for cellular and biochemical in vitro studies.
- High purity (≈98.2%).
- Molecular formula C18H12N2O3S and molecular weight 336.36 g·mol⁻¹.
- CAS number 2285432-57-5 for substance identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.