Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(Benzotriazol-1-ylmethyl)-N-benzyl-1-phenylmethanamine | 57684-32-9 | 95.0% | 328.42 | 1 G
SDP

Supplier:  Medchemexpress LLC HY207921G

Encompass_Preferred

N-(Benzotriazol-1-ylmethyl)-N-benzyl-1-phenylmethanamine is a drug intermediate for the synthesis of various active compounds. This product is intended for research use only.

  • Molecular weight of 328.42
  • Chemical formula C21H20N4
  • CAS number 57684-32-9
  • Appears as a solid
  • Color ranges from white to off-white
  • Purity of 95.0%
  • Documentation available, including data sheet, SDS, COA, and handling instructions

Catalog No. 50-004-01355


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.