Learn More
Medchemexpress LLC N-benzoyl-2'-O-methyladenosine | 85079-00-1 | ≥98.0% | 385.37 | C18H19N5O5 | 5 G

Supplier: Medchemexpress LLC HYW1413945G
Bz-OMe-rA (N-benzoyl-2′-O-methyladenosine) is a nucleoside analog used as a research reagent and antimetabolite probe for studies of nucleoside-related pathways, including cell cycle and DNA damage. It is supplied as a stable material for analytical and cell biology applications, with high purity and defined storage/solubility profiles suitable for laboratory use.
- Chemical name N-benzoyl-2'-O-methyladenosine.
- CAS number 85079-00-1.
- Molecular weight 385.37.
- Molecular formula C18H19N5O5.
- Purity ≥98.0% (by NMR).
- Soluble in DMSO at ≥200 mg/mL.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Intended for use as a nucleoside antimetabolite/analog in research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.