Learn More
Medchemexpress LLC N-benzoyl-2'-O-methyladenosine | 85079-00-1 | MFCD15145188 | ≥98.0% | 385.37 | C18H19N5O5 | 10 G

Supplier: Medchemexpress LLC HYW14139410G
Bz-OMe-rA (N-benzoyl-2'-O-methyladenosine) is a nucleoside analog supplied as a powder for research applications. Manufacturer documents list the compound's CAS, formula, and molecular weight, and provide storage and purity specifications for laboratory use.
- Nucleoside analog used for research applications.
- Powder physical form for handling and storage.
- Purity: ≥98.0% (NMR) as stated in manufacturer certificate of analysis.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Chemical identifiers: CAS 85079-00-1; formula C18H19N5O5; molecular weight 385.37.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.