Learn More
Medchemexpress LLC N-Benzoyl-DL-arginine-4-nitroanilide hydrochloride | 911-77-3 | 99.7% | 434.88 | 500 MG

Supplier: Medchemexpress LLC HYW008953500MG
N-Benzoyl-DL-arginine-4-nitroanilide hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
- High purity: 99.71%
- Solid appearance
- White to light yellow color
- Storage at -20°C, sealed, away from moisture
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.