Learn More
Medchemexpress LLC N-benzoyl-L-aspartic acid | 997815 | MFCD01664832 | 99.9% | 237.21 g/mol | C11H11NO5 | 1 ML

Supplier: Medchemexpress LLC HY11809310MM 1ML
N-benzoyl-L-aspartic acid is a metabolite standard used in research applications, including peptide and protein modification and analytical methods. Supplied forms include solids and a ready-to-use 10 mM solution in DMSO (1 mL). Molecular formula C11H11NO5, molecular weight 237.21 g/mol, CAS 4631-12-3. Intended for research use only.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Available as solid in multiple pack sizes for flexibility.
- High purity (reported 99.89%).
- Molecular formula C11H11NO5 and molecular weight 237.21 g/mol.
- Provided with datasheet and safety data sheet.
- Intended for research and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.