Learn More
Medchemexpress LLC N-Benzoylcytidine | 13089-48-0 | 99.5% | C16H17N3O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY785745G
Encompass_Preferred
N-Benzoylcytidine is a modified cytidine analogue that can be phosphorylated by uracil-cytidine kinase (UCK1 and UCK2). It is intended for research use only.
  • Can be utilized to synthesize 2-OH protective groups for solid-phase RNA synthesis
  • Used for synthetic oligonucleotides for UV induction and targeted gene silencing in zebrafish embryos
Catalog No. 50-004-33326