Learn More
Medchemexpress LLC N-benzyl-N-hydroxy-2,2-dimethylbutanamide | 1956370-21-0 | MFCD30738006 | 99.9% | 221.14 Da | C13H19NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10103210MM 1ML
Encompass_Preferred
RIPA-56 is a potent, selective inhibitor of receptor-interacting protein 1 (RIP1) kinase supplied as a 10 mM solution in DMSO (1 mL) for research use. It is intended for biochemical and cell-based assays to probe RIP1-dependent signaling and cell death pathways. Chemical identifiers include formula C13H19NO2, molecular weight ~221.1-221.3 Da, and CAS 1956370-21-0.
  • Potent RIP1 kinase inhibition (IC50 ≈ 13 nM).
  • Provided as a ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity suitable for research applications.
  • Applicable to biochemical and cell-based assays probing necroptosis and inflammatory signaling.
  • Accompanied by COA and SDS for characterization and safe handling.
Catalog No. 50-002-13590