Learn More
Medchemexpress LLC N-benzyl-N,N-diethylethanaminium iodide | 5400-94-2 | 319.23 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0132315G
Encompass_Preferred
N-Benzyl-N,N-diethylethanaminium iodide is a quaternary ammonium salt composed of a positively charged N-benzyl-N,N-diethylethylamine cation and a negatively charged iodide anion. It is commonly utilized as a phase transfer catalyst in organic chemical reactions, aiding in the transfer of reactants between immiscible phases. Additionally, it can serve as a reagent for synthesizing various organic compounds and as a surfactant or emulsifier in industrial and personal care products. It is intended for research use only.
  • Facilitates the transfer of reactants between immiscible phases in organic chemical reactions.
  • Can be used in the synthesis of various organic compounds.
  • Applicable in industrial and personal care products.
  • Can be used as a biological material or organic compound for life science-related research.
Catalog No. 50-004-54361