Learn More
Medchemexpress LLC N-Benzyl-N N-dimethy 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW105516
Encompass_Preferred
N-Benzyl-N N-dimethylbenzeneminium chloride is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases In addition it is used as an emulsifier surfactant or disinfectant in various industrial applications Its unique chemical properties make it an important ingredient in the production of pharmaceuticals agrochemicals and polymer materials
Catalog No. 50-002-06811