Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Benzylhydroxylamine hydrochloride | 29601-98-7 | MFCD00043377 | 159.61 | 1 KG
SDP

Supplier:  Medchemexpress LLC HYW0022161KG

Encompass_Preferred

N-Benzylhydroxylamine Hydrochloride is a chemical compound that appears as a white to light yellow powder or crystal. It is a solid at 20 °C, suitable for use as biochemical assay reagents, and requires storage under inert gas due to its hygroscopic nature.

  • Solid physical state at 20 °C
  • White to light yellow powder to crystal appearance
  • Recommended storage at room temperature (below 15 °C)
  • Store under inert gas
  • Hygroscopic nature
  • Soluble in DMSO (100 mg/mL with ultrasonic treatment)
  • Causes skin irritation
  • Causes serious eye irritation

Catalog No. 50-004-47380


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.