Learn More
Medchemexpress LLC N-benzylideneaniline | 538-51-2 | MFCD00003027 | 100.0% | 181.23 g/mol | C13H11N | 25 G

Supplier: Medchemexpress LLC 25GHYW025799
N-benzylideneaniline is a small-molecule research reagent reported to inhibit LSD (a histone demethylase). It is supplied as a solid for laboratory biochemical assays and reagent preparation, and is provided with analytical characterization and safety documentation for research use.
- High purity: 99.97%.
- Molecular weight: 181.23 g/mol.
- Chemical formula: C13H11N.
- Available pack sizes: 5 g, 10 g, 25 g, 50 g (quote).
- Includes analytical documents: COA, HNMR, RP-HPLC, and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.