Learn More
Medchemexpress LLC N-((Benzyloxy)carbonyl)-N-methyl-L-valine | 42417-65-2 | 99.46% | 265.30 | 5 G

Supplier: Medchemexpress LLC HYW0117225G
N-((Benzyloxy)carbonyl)-N-methyl-L-valine is a valine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, supplying fuel during exercise, improving mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Storage powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO at 200 mg/mL (753.86 mM); requires ultrasonic assistance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.