Learn More
Medchemexpress LLC N-(Benzyloxycarbonyloxy)succinimide | 13139-17-8 | 99.97% | C12H11NO5 | 1 KG
SDP
Supplier:  Medchemexpress LLC HY752001KG
Encompass_Preferred
N-(Benzyloxycarbonyloxy)succinimide is an effective amino-protecting reagent for organic synthesis. It also functions as a neutral ionophore, which significantly improves the performance of erbium ion (Er3+) sensors, offering a practical tool for rapid detection of erbium ions in environmental and water quality analysis.
  • Efficient amino-protecting reagent
  • Improves erbium ion (Er3+) sensor performance
  • Practical tool for rapid erbium ion detection
Catalog No. 50-004-33688